Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Capacity:50 ml
- Description:Buret with 0,01 ml resolution
- Accuracy:±0,2%
- Pk:1
Specifications
About this item
Digitrate bottle-top burets provide convenient bottle-top digital titration of any reagent.
- Convenient bottle-top digital titration of any reagent
- Constructed of borosilicate glass
- Fluoropolymer resin components
- >0,1% precision
- 0,1% reproducibility
Burets feature large LCD display, safety dispense nozzle protector, and 'zero' reset button for multiple titrations.
Amber, low-profile bottle (sold separately) has a side opening which eliminates the need to remove the Digitrate from the bottle during filling. A safety funnel (also sold separately) has an off-set stem for safer filling of the side neck on the bottle.
Simple user calibration procedure. Precision valve mechanism ensures easy priming with no leakage back into the reservoir. Chemically-resistant liquid pathway. Visual indication of last calibration. Head rotates 360° for use in any position. Burets may be easily dismantled for cleaning without the use of tools. Simple removal of electronics for autoclaving. Digitrate fits directly onto 30 mm neck reservoirs. Each instrument is individually calibrated against National Institute of Standards and Technology Traceable equipment.