To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Temperature range:Ambient +5 to 100 °C
- Description:VWR® Advanced mini block heater with heated lid, TUV listed
- Environmentally Preferable:Waste Reducing
- Environmental certification:RoHS Compliant
- Temperature accuracy:+/- 0,5 °C
- Electrical:120 V, 50/60 Hz, 80 W
- Shipping weight:1,6 kg (3,5 lbs.)
- Temperature uniformity:+/- 0,2 °C
- D×W×H:160×127×127 mm (6.3×5×4.85")
- Pk:1
VWR® Advanced Mini Dry Block Heater with Heated Lid are ideal for a variety of applications that require accurate and repeatable results such as LAMP assays, immunoassays, enzyme reactions, and denaturations.
- Heated lid option reduces condensation on sample lids
- Compact, ergonomic design
- Six modular block options
- Supplied with Best-In-Class Five-Year Warranty
These personal sized heaters take up minimal bench space and are small enough to fit in the palm of your hand.
Block heaters accept one interchangeable mini block for 0,2 ml, 0,5 ml, 1,5 ml, 2 ml, 15 ml, or 50 ml tubes; however, the heated lid model is not designed to accept 15 ml or 50 ml tubes. Both models include a clear polycarbonate lid to help prevent evaporation and maintain temperature uniformity. The cover for the non-heated lid model is easily removable when incubating 15 ml or 50 ml tubes. Efficient heater features close tube and block contact with a low density design for exceptional temperature uniformity. The heated lid model includes a second heater in the lid to help prevent condensation. Units include a block lifting tool. Mini Modular blocks are sold separately.
PID temperature control provides temperature accuracy as low as 0,3 °C and uniformity as low as 0,2 °C. Samples are heated to temperature quickly and accurately. Temperature can be adjusted in 0,1 °C increments. Touch pad controls with easy-to-read, independent LED displays for temperature and time provide repeatable and accurate results every time and are easily visible across lab benches. Timer will display elapsed time, or when programmed to a user defined limit, will shut off when unit reaches zero. Display will show last settings, even after power has been turned off. Temperature calibration mode allows user to calibrate unit to an external temperature device. Cool touch housing is constructed from a high quality, heat and chemical-resistant polymer. The unit’s housing remains cool to the touch throughout normal operating temperatures. In timed mode, an alarm will sound when the timer reaches zero and when the set temperature is reached. The heated lid helps to regulate the temperature and reduce the amount of condensation on sample lids.
NOTE: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Multiple data points within the temperature range and time function are reported on the certificate with the associated uncertainties. All units include a 234 cm (92") detachable, three-wire cord and plug. Units come with a five-year warranty on parts and labor. 230 V, 50/60 Hz models are also available; contact your Avantor sales representative for more information.