illustra™ plasmidPrep Mini Spin kit
About this item
The illustra™ plasmidPrep Mini Spin Kit uses a simple plasmid DNA purification protocol involving a modified alkaline lysis procedure and a novel silica-based membrane to achieve highly efficient plasmid DNA purification. Produces high quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing.
- Produces high-quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing
- Total preparation time of less than 10 min represents a 50% reduction in protocol time
- Increased ease of use provided by minimal changes in pipetting volume from one part of the protocol to the next
- Fewer centrifugation steps
- Color-coded tubes and columns simplify the protocol
- No organic solvents such as phenol chloroform
The extracted plasmid DNA resulting from performing a miniprep is called a "plasmid miniprep" and is often performed using a plasmid purification kit. Minipreps are used in the process of molecular cloning to analyze bacterial clones. When bacteria are lysed under alkaline conditions, both chromosomal DNA and protein are denatured, but the plasmid DNA remains stable. On addition of acetate-containing neutralization buffer the larger and less supercoiled genomic DNA and proteins precipitate, but the small bacterial DNA plasmids stay in the solution.
The typical plasmid DNA yield from a fresh 1.5 mL culture of E. coli containing a high copy number plasmid is approximately 9 μg. Instead of organic solvents, the kit contains chaotropic salts to denature protein components and promote selective binding of DNA to the novel silica membrane.
Delivery information: The kit contains spin columns prepacked with a novel silica membrane, suspension, lysis, and neutralization buffers containing chaotropic salts, wash and elution buffers, microcentrifuge collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.
2 Options Available Below
Product Details & Documents
The illustra™ plasmidPrep Mini Spin Kit uses a simple plasmid DNA purification protocol involving a modified alkaline lysis procedure and a novel silica-based membrane to achieve highly efficient plasmid DNA purification. Produces high quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing.
- Produces high-quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing
- Total preparation time of less than 10 min represents a 50% reduction in protocol time
- Increased ease of use provided by minimal changes in pipetting volume from one part of the protocol to the next
- Fewer centrifugation steps
- Color-coded tubes and columns simplify the protocol
- No organic solvents such as phenol chloroform
The extracted plasmid DNA resulting from performing a miniprep is called a "plasmid miniprep" and is often performed using a plasmid purification kit. Minipreps are used in the process of molecular cloning to analyze bacterial clones. When bacteria are lysed under alkaline conditions, both chromosomal DNA and protein are denatured, but the plasmid DNA remains stable. On addition of acetate-containing neutralization buffer the larger and less supercoiled genomic DNA and proteins precipitate, but the small bacterial DNA plasmids stay in the solution.
The typical plasmid DNA yield from a fresh 1.5 mL culture of E. coli containing a high copy number plasmid is approximately 9 μg. Instead of organic solvents, the kit contains chaotropic salts to denature protein components and promote selective binding of DNA to the novel silica membrane.
Delivery information: The kit contains spin columns prepacked with a novel silica membrane, suspension, lysis, and neutralization buffers containing chaotropic salts, wash and elution buffers, microcentrifuge collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- GEHE28-9042-69
- GEHE28-9042-70
- Description:
- illustra™ plasmidPrep Mini Spin Kit, 50 preps
- illustra™ plasmidPrep Mini Spin Kit, 250 preps
- Target molecule:
- Plasmid
- Plasmid
- Refine target molecule:
- Plasmid
- Plasmid
- Sample type:
- Bacteria
- Bacteria
- Method / Format:
- Column (silica-based)
- Column (silica-based)
- Prep size:
- Mini
- Mini
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- GEHE28-9042-69illustra™ plasmidPrep Mini Spin Kit, 50 prepsSpecifications:
More
Specifications
Cat. No.GEHE28-9042-69Descriptionillustra™ plasmidPrep Mini Spin Kit, 50 prepsTarget moleculePlasmidRefine target moleculePlasmidSample typeBacteriaMethod / FormatColumn (silica-based)Prep sizeMini - GEHE28-9042-70illustra™ plasmidPrep Mini Spin Kit, 250 prepsSpecifications:
More
Specifications
Cat. No.GEHE28-9042-70Descriptionillustra™ plasmidPrep Mini Spin Kit, 250 prepsTarget moleculePlasmidRefine target moleculePlasmidSample typeBacteriaMethod / FormatColumn (silica-based)Prep sizeMini
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



