



Hexane (mixture of isomers) ≥95% (by GC, sum of isomers + methylcyclopentane), J.T.Baker®
Catalog # BAKRN169-08
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC):≥95 %
- Identification (by IR):Passes Test
- Appearance (Clear, colorless liquid):Passes Test
- Color (APHA):≤10
- Residue after Evaporation:≤20 ppm
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:68…70 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:Management of Change (MOC) category = N
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311