Order Entry
China
ContactUsLinkComponent
EDTA disodium salt 0.0575 M in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
EDTA disodium salt 0.0575 M in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MACRH109-05
CAS Number:  
EDTA disodium salt 0.0575 M in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MACRH109-05
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Molarity:
    0.0570 - 0.0580

Specification Test Results

About this item

 :
 :336.21 g/mol
 :Management of Change (MOC) category = NI
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672