Order Entry
China
ContactUsLinkComponent
Sulphuric acid 0.125 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sulphuric acid 0.125 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
目录编号 BAKR5692-03
Supplier:  Avantor
CAS Number:  
Sulphuric acid 0.125 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
目录编号 BAKR5692-03
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Cubetainer®
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Normality:
    0.1245 - 0.1255
  • Chloride (Cl):
    ≤1 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Heavy Metals (as Pb):
    ≤0.3 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

Specification Test Results

About this item

 :
 :98.08 g/mol
ADR:8,II
 :Management of Change (MOC) category = NI
UN:1830
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589