Order Entry
China
Orders LinkContactUsLinkComponent
Bromine 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Bromine 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Poly bottle, 1 L
Catalog # BAKR5625-02
Health
Supplier:   Avantor
Bromine 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # BAKR5625-02
Supplier:  Avantor
CAS Number:  7726-95-6
Bromine 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Poly bottle, 1 L

CAS Number:  7726-95-6
Price
Inquire For Price
Log in for availability
1 * 1 升
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7726-95-6
  • Formula:Br₂
  • MDL Number:MFCD00010896
  • Molecular Weight:159.81 g/mol
  •  :Management of Change (MOC) category = N

Documents

Specifications

Specification Test Results

  • Normality:
    0.095 - 0.105

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.