


Acetic acid glacial ≥99.7% (by freezing point), CMOS for microelectronic, J.T.Baker®
Catalog # BAKR9502-89
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Assay (CH₃COOH) (by freezing point):≥99.7 %
- Color (APHA):≤10
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Acetaldehyde:≤0.05 %
- Residue after Evaporation:≤4 ppm
- Solubility in H₂O:Passes Test
- Specific Gravity at 20°/20°C:≥1.048
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤1 ppm
- Phosphate (PO₄):≤1 ppm
- Sulfate (SO₄):≤0.5 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Arsenic and Antimony (as As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤50 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤30.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤20.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Heavy Metals (as Pb):≤300.0 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤25.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silicon (Si):≤50 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤100.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤50.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤100.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
Specification Test Results
About this item
:
:60.05 g/mol
Melting Pt:16.4 °C
Storage Temperature:Ambient
UN:2789
ADR:8,II
:GHS02
:GHS05
:GHS07
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152