Order Entry
China
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Biotech Reagent, J.T.Baker®
Irritant
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Biotech Reagent, J.T.Baker®
Catalog # BAKR4126-07
Supplier:  Avantor
CAS Number:  
Irritant
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Biotech Reagent, J.T.Baker®
Catalog # BAKR4126-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Assay (NH₂C(CH₂OH)₃·HCl):
    99.0 - 101.0 %
  • Appearance:
    Passes Test
  • Absorbance of a 1M Solution - 260 nm:
    ≤0.06
  • Absorbance of a 1M Solution - 280 nm:
    ≤0.06
  • Absorbance of a 1M Solution - 400 nm:
    ≤0.01
  • Endotoxin Concentration (EU/g):
    ≤2.5
  • Identification (by IR):
    Passes Test
  • Melting Point:
    147 - 153 °C
  • pH of 0.5M Solution at 25°C:
    3.5 - 5.0
  • Loss on Drying at 105°C:
    ≤0.5 %
  • Trace Impurities - Arsenic (As):
    ≤10 ppm
  • Trace Impurities - Calcium (Ca):
    ≤10.0 ppm
  • Trace Impurities - Copper (Cu):
    ≤10 ppm
  • Trace Impurities - Iron (Fe):
    ≤10 ppm
  • Trace Impurities - Lead (Pb):
    ≤10 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤10 ppm

Specification Test Results

About this item

 :
 :157.6 g/mol
 :GHS07
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590