Avantor® ACE® UltraCore BIO, Analytical HPLC/UHPLC Columns
Avantor® ACE® UltraCore BIO C18 are high performing solid-core (core-shell) particle columns designed for a wide range of biomolecule applications. With both 300 Å and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of more hydrophilic proteins over 5 kDa molecular weight.
- Wide pore, solid-core combination for excellent peak shape & resolution of large molecules
- High purity, base deactivated silica with excellent reproducibility
- Good choice for fast analysis of polypeptides
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
- Catalog No:
- ACEUBIO-250-1030
- ACEUBIO-350-0546
- ACEUBIO-350-546G
- ACEUBIO-350-0521
- ACEUBIO-350-1521
- ACEUBIO-350-1021
- ACEUBIO-250-1530
- ACEUBIO-250-521G
- ACEUBIO-250-546G
- ACEUBIO-250-0521
- ACEUBIO-350-1046
- ACEUBIO-350-1546
- ACEUBIO-350-530G
- ACEUBIO-350-0530
- ACEUBIO-250-1046
- ACEUBIO-250-0546
- ACEUBIO-250-1021
- ACEUBIO-250-1546
- ACEUBIO-250-530G
- ACEUBIO-250-1521
- ACEUBIO-350-1030
- ACEUBIO-250-0530
- ACEUBIO-350-521G
- ACEUBIO-350-1530
- Description:
- 500 Å
- 300 Å
- For analytical columns with 4,6 mm Int.Ø
- 300 Å
- 300 Å
- 300 Å
- 500 Å
- For analytical columns with 2,1 mm Int.Ø
- For analytical columns with 4,6 mm Int.Ø
- 500 Å
- 300 Å
- 300 Å
- For analytical columns with 3,0 mm Int.Ø
- 300 Å
- 500 Å
- 500 Å
- 500 Å
- 500 Å
- For analytical columns with 3,0 mm Int.Ø
- 500 Å
- 300 Å
- 500 Å
- For analytical columns with 2,1 mm Int.Ø
- 300 Å
- Carbon load:
- 1,4%
- 1,0%
- 1,0%
- 1,0%
- 1,0%
- 1,0%
- 1,4%
- 1,4%
- 1,4%
- 1,4%
- 1,0%
- 1,0%
- 1,0%
- 1,0%
- 1,4%
- 1,4%
- 1,4%
- 1,4%
- 1,4%
- 1,4%
- 1,0%
- 1,4%
- 1,0%
- 1,0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Surface area:
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 16 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- Temperature limit:
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1