
Tetrahydro-4H-pyran-4-one 99%
ACRO189440050
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4530 (20°C, 589 nm)
- GC:≥98.5 %
:
:100.12 g/mol
:166 °C (760 mmHg)
:1.08 g/cm³
:56 °C
:Ambient
:249-967-2
:1224
:3,III
:GHS02
:29943-42-8
:C₅H₈O₂
:MFCD00006581