


Ammonium metavanadate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7803-55-6
:
Formula:NH₄VO₃
MDL Number:MFCD00011430
:116.98 g/mol
Merck Index:13,00572
Melting Pt:200 °C (dec.)
Density:2,33 g/cm³ (20 °C)
EINECS:232-261-3
UN:2859
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow to light green Crystalline powder
- Titration with KMnO4:≥99.4 %
- Chloride (Cl):≤20 ppm
- Phosphate (PO4):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Cadmium (Cd):≤10 ppm
- Cobalt (Co):≤20 ppm
- Copper (Cu):≤10 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤20 ppm
- Nickel (Ni):≤20 ppm
- Zinc (Zn):≤10 ppm