
Ammonium hexafluorotitanate 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16962-40-6
:
Formula:H₈F₆N₂Ti
MDL Number:MFCD00010888
:197.95 g/mol
Storage Temperature:Ambient
EINECS:241-036-9
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Total trace metal impurities:≤0.01 %