


Lithium hexafluorophosphate 98%, pure
ACRO191260250
:
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Appearance:White to off-white Powder
- Assay:≥97.5 % (ex Li)
:
:~ 1,5 g/cm³ (20 °C)
:244-334-7
:8,I
:151.91 g/mol
:2923
:GHS05
:GHS06
:GHS08
:Ambient
:21324-40-3
:F₆LiP
:MFCD00011096