Order Entry
China
ContactUsLinkComponent
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole ≥97.0% (by HPLC)
刺激性
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole ≥97.0% (by HPLC)
Catalog # TCIAT2624-1G
供应商:  TCI
CAS 编号:  
刺激性
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole ≥97.0% (by HPLC)
Catalog # TCIAT2624-1G
供应商:  TCI
CAS 编号:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

规格

  • Pk:
    1 g
  • Seq:
    3047

规格

规格测试结果

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Melting point:
    76.0 to 80.0 deg-C

规格测试结果

关于此项目

 :
 :330.09 g/mol
 :GHS07
储存温度:Refrigerator
CAS 编号:887144-97-0
化学式:C₁₀H₁₀F₃IO
MDL 编号:MFCD10567056