ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥90.0% (by HPLC) for biochemical research
Catalog # TCIAT0521-100MG
Supplier: TCI
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 90.0 area%
- abs-ratio (a/b):0.41 to 0.44 (E340/E260, pH10.0)
- Molar extinction coefficient:min. 12900 (pH10.0, 260 nm)
- Molar extinction coefficient2:min. 5400 (pH10.0, 340 nm)
Specification Test Results
About this item
:
:833.35 g/mol
Merck Index:14,06348
Storage Temperature:Freezer
EINECS:220-163-3
CAS Number:2646-71-1
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number:MFCD00036263