Order Entry
China
ContactUsLinkComponent
β-Naphthol violet
β-Naphthol violet
Catalog # TCIAN0037-1G
Supplier:  TCI
CAS Number:  
β-Naphthol violet
Catalog # TCIAN0037-1G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark red to Brown powder to crystal
  • Lambda max.:
    508.0 to 511.0nm (H2O)
  • Absorbance (E1%1cm):
    min. 520 (5mg/L, H2O, 508.0 to 511.0nm)
  • Drying loss:
    max. 10.0 %
  • Sensitiveness:
    to pass test

Specification Test Results

About this item

 :
 :497.36 g/mol
EINECS:230-443-7
CAS Number:7143-21-7
Formula:C₁₆H₁₁N₃O₉S₂
MDL Number:MFCD00021457