Order Entry
China
ContactUsLinkComponent
trans-β-Methylstyrene ≥97.0% (by GC) stabilised
Flammable
Irritant
Environment
trans-β-Methylstyrene ≥97.0% (by GC) stabilised
Catalog # TCIAM1175-10ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
Environment
trans-β-Methylstyrene ≥97.0% (by GC) stabilised
Catalog # TCIAM1175-10ML
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :118.18 g/mol
Boiling Pt:176°C
Melting Pt:-29°C
Melting Pt:–29°C
Flash Pt:52°C
Storage Temperature:Refrigerator
EINECS:212-848-0
UN:3295
ADR:3,III
 :Stabilised with 4-tert-butylpyrocatechol (TBC)
 :GHS02
 :GHS07
 :GHS09
CAS Number:873-66-5
Formula:C₉H₁₀
MDL Number:MFCD00009280