订单输入
China
ContactUsLinkComponent
6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one ≥98.0% (通过滴定分析)
6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one ≥98.0% (通过滴定分析)
Catalog # TCIAM1116-5G
Supplier:  TCI
CAS Number:  
6-Methyl-2H-pyrido[1,2-a]pyrimidin-2-one ≥98.0% (通过滴定分析)
Catalog # TCIAM1116-5G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • 包装规格:
    5 g
  • 顺序:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :160.18 g/mol
CAS Number:16075-68-6
Formula:C₉H₈N₂O
MDL Number:MFCD00191466