Order Entry
China
ContactUsLinkComponent
3-Methyl-1-(4-sulphophenyl)-2-pyrazolin-5-one ≥97.0% (by titrimetric analysis)
Corrosive
3-Methyl-1-(4-sulphophenyl)-2-pyrazolin-5-one ≥97.0% (by titrimetric analysis)
Catalog # TCIAM0090-25G
Supplier:  TCI
CAS Number:  
Corrosive
3-Methyl-1-(4-sulphophenyl)-2-pyrazolin-5-one ≥97.0% (by titrimetric analysis)
Catalog # TCIAM0090-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :254.26 g/mol
EINECS:201-901-3
UN:1759
ADR:8,II
 :GHS05
CAS Number:89-36-1
Formula:C₁₀H₁₀N₂O₄S
MDL Number:MFCD00020756