Order Entry
China
ContactUsLinkComponent
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one ≥98.0% (by GC)
Irritant
Environment
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one ≥98.0% (by GC)
Catalog # TCIAD4157-25G
Supplier:  TCI
CAS Number:  
Irritant
Environment
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one ≥98.0% (by GC)
Catalog # TCIAD4157-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    40.0 to 44.0 deg-C

Specification Test Results

About this item

 :
Merck Index:14,03070
ADR:9,III
 :282.22 g/mol
EINECS:264-843-8
UN:3077
 :GHS07
 :GHS09
CAS Number:64359-81-5
Formula:C₁₁H₁₇Cl₂NOS
MDL Number:MFCD04034673