Order Entry
China
ContactUsLinkComponent
2,3-Dihydro-2-spiro-4'-[8'-aminonaphthalen-1'(4'H)-one]perimidine ≥98.0% (by total nitrogen basis) (contains o-form)
Irritant
2,3-Dihydro-2-spiro-4'-[8'-aminonaphthalen-1'(4'H)-one]perimidine ≥98.0% (by total nitrogen basis) (contains o-form)
Catalog # TCIAD3618-100MG
Supplier:  TCI
CAS Number:  
Irritant
2,3-Dihydro-2-spiro-4'-[8'-aminonaphthalen-1'(4'H)-one]perimidine ≥98.0% (by total nitrogen basis) (contains o-form)
Catalog # TCIAD3618-100MG
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

About this item

 :
 :313.36 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:851768-63-3
Formula:C₂₀H₁₅N₃O