Order Entry
China
ContactUsLinkComponent
1,1-Diphenyl-4-(piperidin-1-yl)butan-1-ol hydrochloride ≥99.0% (by titrimetric analysis)
刺激性
1,1-Diphenyl-4-(piperidin-1-yl)butan-1-ol hydrochloride ≥99.0% (by titrimetric analysis)
Catalog # TCIAD2062-25G
供应商:  TCI
CAS 编号:  
刺激性
1,1-Diphenyl-4-(piperidin-1-yl)butan-1-ol hydrochloride ≥99.0% (by titrimetric analysis)
Catalog # TCIAD2062-25G
供应商:  TCI
CAS 编号:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

规格

  • Pk:
    25 g
  • Seq:
    3308

规格

规格测试结果

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Solubility in Methanol:
    almost transparency

规格测试结果

关于此项目

 :
 :345.91 g/mol
默克索引:14,03311
EINECS:221-850-0
 :GHS07
CAS 编号:3254-89-5
化学式:C₂₁H₂₈ClNO
MDL 编号:MFCD00151479