Order Entry
China
ContactUsLinkComponent
Copper(II) dimethyldithiocarbamate ≥98.0% (by titrimetric analysis)
Environment
Copper(II) dimethyldithiocarbamate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1022-25G
Supplier:  TCI
CAS Number:  
Environment
Copper(II) dimethyldithiocarbamate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1022-25G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :303.96 g/mol
EINECS:205-287-8
UN:3077
ADR:9,III
 :GHS09
CAS Number:137-29-1
Formula:C₃H₇NS₂
MDL Number:MFCD00050845