Order Entry
China
ContactUsLinkComponent
Ceric ammonium molybdate solution (contains H₂SO₄) stain, for TLC
Corrosive
Health
Ceric ammonium molybdate solution (contains H₂SO₄) stain, for TLC
Catalog # TCIAC1794-500ML
Supplier:  TCI
CAS Number:  
Corrosive
Health
Ceric ammonium molybdate solution (contains H₂SO₄) stain, for TLC
Catalog # TCIAC1794-500ML
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Yellow to Amber clear liquid
  • Functionality test (TLC):
    A blue spot appears on 4-Methoxyphenol
  • Sensitiveness:
    A blue spot appears on 4-Methoxyphenol

Specification Test Results

About this item

 :
UN:3264
ADR:8,III
 :GHS05
 :GHS08
MDL Number:MFCD06797081