Order Entry
China
ContactUsLinkComponent
Lithium acetate, anhydrous 99+% for analysis
Irritant
Lithium acetate, anhydrous 99+% for analysis
Catalog # ACRO297110250
CAS Number:  
undefined
Irritant
Lithium acetate, anhydrous 99+% for analysis
Catalog # ACRO297110250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    280 to 285 °C
  • Water:
    ≤0.3 % (K.F.)
  • Titration with HClO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤200 ppm
  • Calcium (Ca):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Magnesium (Mg):
    ≤10 ppm
  • Potassium (K):
    ≤50 ppm
  • Sodium (Na):
    ≤100 ppm
  • Heavy metals (as Pb):
    ≤20 ppm

Specification Test Results

About this item

 :
Merck Index:12,05544
Melting Pt:280…285 °C
Density:1,26 g/cm³ (20 °C)
 :65.99 g/mol
EINECS:208-914-3
 :GHS07
Storage Temperature:Ambient
CAS Number:546-89-4
Formula:H₃CCOOLi
MDL Number:MFCD00013057