Order Entry
China
ContactUsLinkComponent
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride hydrochloride ≥97.0% (by titrimetric analysis) for HPLC labeling
Corrosive
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride hydrochloride ≥97.0% (by titrimetric analysis) for HPLC labeling
Catalog # TCIAA5579-100MG
Supplier:  TCI
CAS Number:  
undefined
Corrosive
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride hydrochloride ≥97.0% (by titrimetric analysis) for HPLC labeling
Catalog # TCIAA5579-100MG
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Effect test of HPLC derivatization:
    Effective as labeling agent for Phentermine

Specification Test Results

About this item

 :
 :395.28 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
 :GHS05
Formula:C₂₂H₁₅ClN₂O
MDL Number:MFCD03844764