Order Entry
China
ContactUsLinkComponent
Acid red 52 ≥98.0% (by HPLC)
Acid red 52 ≥98.0% (by HPLC)
Catalog # TCIAA0600-25G
Supplier:  TCI
CAS Number:  
undefined
Acid red 52 ≥98.0% (by HPLC)
Catalog # TCIAA0600-25G
Supplier:  TCI
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Red to Dark blue to Black powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Lambda max.:
    564.0 to 568.0 nm (1.5g/L AcONH4 sol.)
  • Absorbance (E1%1cm):
    min. 1,800 (1.5g/L AcONH4 sol., 564.0 to 568.0 nm)

Specification Test Results

About this item

Synonyms: Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200
 :
 :580.65 g/mol
EINECS:222-529-8
CAS Number:3520-42-1
Formula:C₂₇H₃₀N₂O₇S₂
MDL Number:MFCD00010180