Order Entry
China
ContactUsLinkComponent
Sodium sulphide hydrate, GPR RECTAPUR®
Toxic
Corrosive
Environment
Sodium sulphide hydrate, GPR RECTAPUR®
Catalog # VWRC83757.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Environment
Sodium sulphide hydrate, GPR RECTAPUR®
Catalog # VWRC83757.290
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    58 - 64 %
  • SO3 (Sulphite):
    Max. 0.6 %
  • S2O3 (Thiosulphate):
    Max. 0.5 %

Specification Test Results

About this item

 :
 :96.06 g/mol (anhy)
Merck Index:12,08829
Boiling Pt:~920 °C (1013 hPa)
Melting Pt:50 °C
Density:∼1.43 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:215-211-5
UN:1849
ADR:8,II
REACH:01-2119513694-38
 :GHS06
 :GHS05
 :GHS09
CAS Number:27610-45-3
Formula:Na₂S·~3H₂O
MDL Number:MFCD00149183