Order Entry
China
ContactUsLinkComponent
L(-)-Cystine 98.5-101.5% (dried basis) FCC for biopharmaceutical production, SAFC®
L(-)-Cystine 98.5-101.5% (dried basis) FCC for biopharmaceutical production, SAFC®
  MERP1.02836.1000
 :  
L(-)-Cystine 98.5-101.5% (dried basis) FCC for biopharmaceutical production, SAFC®
  MERP1.02836.1000
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

 

 

  • Assay (ex N, calc. on dried substance):
    98.5 - 101.5 %
  • Identity (IR-spectrum):
    Passes Test
  • Appearance:
    White to almost white, fine-crystalline powder
  • Appearance of solution (100 g/l, hydrochloric acid 2 mol/l):
    Clear and Colorless
  • Spec. Rotation (α20/D, 20 g/l, Hydrochloric Acid 1 mol/l, Previously Dried Substance):
    –215 to –225
  • Heavy Metals (as Pb):
    ≤10 ppm
  • Pb (Lead):
    ≤5 ppm
  • Ninhydrin - Positive Substances (LC) (Largest Single Impurity):
    ≤0.2 %
  • Ninhydrin - Positive Substances (LC) (Sum of all Impurities):
    ≤0.5 %
  • Sulfated Ash (800 °C):
    ≤0.1 %
  • Loss on Drying (105 °C; 3 h):
    ≤0.2 %

 

 

 : (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
 :
 :240.3 g/mol
 :13,02811
 :428 °C (1013 hPa)
 :261…262 °C (dec.)
 :1.677 g/cm³ (20 °C)
 :Ambient
 :200-296-3
 :56-89-3
 :HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
 :MFCD00064228