Sodium dihydrogen phosphate monohydrate 99.0-102.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
: Avantor
:10049-21-5
:
:NaH₂PO₄·H₂O
:MFCD00149208
:137.99 g/mol
:13,08734
:100 °C
:2.04 g/cm³ (20 °C)
:Ambient
:231-449-2
:01-2119489796-13
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- For Laboratory,Research,or Manufacturing Use:99.0 - 102.0 %
- Assay (NaH₂PO₄·H₂O):99.0 - 102.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- pH of 5% Solution at 25°C:4.1 - 4.5
- Insoluble Matter, Calcium and NH₄OH Precipitate:≤0.01 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %