Sodium hypophosphite monohydrate 99-103% (by titrimetric analysis) for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10039-56-2
:
Formula:NaH₂PO₂·H₂O
MDL Number:MFCD06252379
:105.99 g/mol
Melting Pt:90 °C
Density:0,8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-669-9
Specification Test Results
- Appearance:White Powder or crystals
- pH:6 to 8 (5 % in water at 20°C)
- Titration:99.0 to 103.0 %
- Chloride (Cl):≤200 ppm
- Arsenic (As):≤2 ppm
- Calcium (Ca):≤200 ppm
- Iron (Fe):≤5 ppm
- Lead (Pb):≤5 ppm