

Dodecamolybdophosphoric acid hydrate ACS
:51429-74-4
:
:H₃[P(Mo₃O₁₀)₄]·nH₂O
:MFCD00011341
:1843.27 g/mol
:12,07498
:78 - 90 °C
:1,62 g/cm³ (25 °C)
:Ambient
:234-713-5
:3084
:8,II
:GHS05
:GHS03
- Appearance:Yellow to yellow-orange Adhering crystals
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤250 ppm
- Ammonium (NH4):≤100 ppm
- Calcium (Ca):≤200 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤50 ppm
- Insoluble matter:≤0.01 %