

Dodecamolybdophosphoric acid hydrate 80%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51429-74-4
:
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913
:1843.27 g/mol
Merck Index:12,07498
Melting Pt:78…90 °C
Density:1,62 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-713-5
UN:3084
ADR:8,II
:GHS05
:GHS03
Specification Test Results
- Appearance:Yellow to yellow-orange Adhering crystals
- Assay:≥47.5 % (Mo)
- Loss on drying:≤25 %
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤250 ppm
- Ammonium (NH4):≤100 ppm
- Calcium (Ca):≤200 ppm
- Iron (Fe):≤50 ppm