Order Entry
China
ContactUsLinkComponent
VWR® ARISTAR®, Ytterbium Standard Solution, 1000 mg/L Yb in dil. Nitric Acid (from Yb₂O₃), Standard for ICP
VWR® ARISTAR®, Ytterbium Standard Solution, 1000 mg/L Yb in dil. Nitric Acid (from Yb₂O₃), Standard for ICP
Catalog # VWRC456344C
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Ytterbium Standard Solution, 1000 mg/L Yb in dil. Nitric Acid (from Yb₂O₃), Standard for ICP
Catalog # VWRC456344C
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Ytterbium
  • Symbol:
    Yb

Specifications

About this item

Yb₂O₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.

Supplied with Certificate of Analysis.