2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent
Catalog # VWRC23347.105
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- IR Spectrum:Passes test
- Interfering dyestuffs:Passes test ACS
- Loss on drying (120°C):Max. 12 %
- Conforms to ACS:Passes test
Specification Test Results
About this item
:
:290.08 g/mol (anhy)
Melting Pt:> 300 °C
EINECS:210-640-4
CAS Number:1266615-56-8
:620-45-1
Formula:C₁₂H₆Cl₂NNaO₂·xH₂O
MDL Number:MFCD00150014