Order Entry
China
ContactUsLinkComponent
Indigo carmine analytical reagent
Irritant
Indigo carmine analytical reagent
Catalog # VWRC22537.138
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Indigo carmine analytical reagent
Catalog # VWRC22537.138
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :466.36 g/mol
Merck Index:13,04969
Melting Pt:> 300 °C
EINECS:212-728-8
 :GHS07
Storage Temperature:Ambient
CAS Number:860-22-0
Formula:C₁₆H₈N₂Na₂O₈S₂
MDL Number:MFCD00005723