Order Entry
China
ContactUsLinkComponent
EDTA dipotassium salt dihydrate ≥97.0% analytical reagent
EDTA dipotassium salt dihydrate ≥97.0% analytical reagent
Catalog # VWRC20490.188
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA dipotassium salt dihydrate ≥97.0% analytical reagent
Catalog # VWRC20490.188
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 97.0 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 0.05 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :404.46 g/mol
Merck Index:13,03546
Melting Pt:272 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:217-895-0
CAS Number:25102-12-9
Formula:C₁₀H₁₄K₂N₂O₈·2H₂O
MDL Number:MFCD00150036