Order Entry
China
ContactUsLinkComponent
 
Flammable
Health
Irritant
Ethanol denatured 90% ≥90%, GPR RECTAPUR® (denatured with methanol)
Catalog # VWRC30243EM
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Ethanol denatured 90% ≥90%, GPR RECTAPUR® (denatured with methanol)
Catalog # VWRC30243EM
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic jerrican, non-removable head
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Density (20/4):
    0.811 0.816
  • Free acidity:
    Max. 0.001 meq/g

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:78.3 °C (1013 hPa)
Flash Pt:12 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568