Order Entry
China
Orders LinkContactUsLinkComponent
 

VWR®, General purpose viscosity standards

VWR®, General purpose viscosity standards
Supplier:  VWR Chemicals
Certificates
VWR®, General purpose viscosity standards
VWR®, General purpose viscosity standards

28 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These ISO 17025 and ISO 17034 certified general purpose viscosity standards provide certified kinematic viscosity, dynamic viscosity and density data across temperature ranges from 20 to 100 °C.

  • Certified in strict accordance with ASTM D2162
  • Density g/ml in accordance with ASTM D1480
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) and dynamic viscosity (mPa·s) given at all temperatures
  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging

Certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture.

These products have been tested at primary level using master viscometers in accordance with ASTM D2162 to ensure the lowest uncertainty of measurement, thus providing standards that are certified with primary level accuracy. ASTM D2162 is the only referenced method for the certification of viscosity standards when adopting ASTM test method protocols.

 : Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.

 : The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.

Product Family Options

Product Information

  • No.
    Viscosity at 25 °C
    Pack type
    Test method
    Pk
    Availability
    Price
  • VWRC99456.260
    Standard with certified values at 20 °C to 60 °C
    Standard with certified values at 20 °C to 60 °C
    N2
    2,144 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,475.00
    ¥2,475.00
    1 * 500 毫升
       ¥2,475.00

  • VWRC99472.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S3
    3,291 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99474.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S6
    7,442 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99466.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N7.5
    8,559 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99469.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S20
    29,04 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,481.00
    ¥2,481.00
    1 * 500 毫升
       ¥2,481.00

  • VWRC99459.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N26
    48,59 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,659.00
    ¥2,659.00
    1 * 500 毫升
       ¥2,659.00

  • VWRC99460.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N35
    55,36 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,679.00
    ¥2,679.00
    1 * 500 毫升
       ¥2,679.00

  • VWRC99462.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N44
    71,11 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99475.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S60
    100,7 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,696.00
    ¥2,696.00
    1 * 500 毫升
       ¥2,696.00

  • VWRC99467.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N75
    124,9 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,627.00
    ¥2,627.00
    1 * 500 毫升
       ¥2,627.00

  • VWRC99451.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N140
    258 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,455.00
    ¥2,455.00
    1 * 500 毫升
       ¥2,455.00

  • VWRC99470.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S200
    392 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,692.00
    ¥2,692.00
    1 * 500 毫升
       ¥2,692.00

  • VWRC99457.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N250
    488,1 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥1,709.00
    ¥1,709.00
    1 * 500 毫升
       ¥1,709.00

  • VWRC99461.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N350
    717,4 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,708.00
    ¥2,708.00
    1 * 500 毫升
       ¥2,708.00

  • VWRC99463.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N415
    796,5 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,476.00
    ¥2,476.00
    1 * 500 毫升
       ¥2,476.00

  • VWRC99476.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S600
    1273 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,622.00
    ¥2,622.00
    1 * 500 毫升
       ¥2,622.00

  • VWRC99468.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N750
    1690 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,629.00
    ¥2,629.00
    1 * 500 毫升
       ¥2,629.00

  • VWRC99452.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N1000
    2610 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,629.00
    ¥2,629.00
    1 * 500 毫升
       ¥2,629.00

  • VWRC99453.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N1400
    3481 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99471.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S2000
    4599 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,718.00
    ¥2,718.00
    1 * 500 毫升
       ¥2,718.00

  • VWRC99458.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N2500
    6618 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99464.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N4000
    10030 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,489.00
    ¥2,489.00
    1 * 500 毫升
       ¥2,489.00

  • VWRC99465.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N5100
    15406 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥1,709.00
    ¥1,709.00
    1 * 500 毫升
       ¥1,709.00

  • VWRC99477.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S8000
    19588 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,626.00
    ¥2,626.00
    1 * 500 毫升
       ¥2,626.00

  • VWRC99450.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N10200
    30991 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥1,709.00
    ¥1,709.00
    1 * 500 毫升
       ¥1,709.00

  • VWRC99454.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N15000
    40049 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • VWRC99455.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    N18000
    54900 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升 - ¥2,629.00
    ¥2,629.00
    1 * 500 毫升
       ¥2,629.00

  • VWRC99473.260
    Standard with certified values at 20 °C to 100 °C
    Standard with certified values at 20 °C to 100 °C
    S30000
    72328 mPa·s
    Glass bottle
    ASTM D2162
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.