About this item
DEC-CYCLE® II is a low pH phenolic disinfectant that is effective as hard non-porous surface disinfectant/detergent formulated for effective aseptic and controlled environment sanitation. When used as directed, DEC-CYCLE II’s sterile pharmaceutical cleanroom formula is for broad spectrum disinfection use on numerous surfaces that require cleaning. Acceptable for use by pharmaceutical and biotechnology industries, medical device manufacturers, hospitals, and any health care institutions.
- Filled in an ISO 5 (Grade A/B, Former Class 100) and filtered at 0.2 microns
- Sterile pharmaceutical cleanroom formula
- Components are air washed with 0.2 micron filtered air before assembly
- Gamma irradiated at a 10-6 SAL
- Lot sterility tested according to current USP compendium
- Completely traceable from start to finish
- Completely validated for sterility and shelf life
Broad spectrum efficacy confirmed by using A.O.A.C protocol testing at a use dilution of 1:256.
Effective against gram negative Pseudomonas aeruginosa and gram positive Staphylococcus aureus bacteria.
Applications include: On hard, inanimate surfaces in aseptic filling and gowning rooms, and general manufacturing areas, on machinery, tables, counters, laminar flow benches, floors, and walls with a range of materials including stainless steel, glazed porcelain, glass, and chrome
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.