Methyl myristate, standard material for GC
Catalog # TCIAS0310-5ML
Supplier: TCI
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:124-10-7
- Description:Methyl Myristate, Purity (GC): min. 99.5 %
- Formula:C₁₅H₃₀O₂
- Description:Methyl Myristate
- Flash point:"157 °C (lit.)"
- Boiling point:173 °C/20 mmHg (lit.)
- Freezing point:"18 °C"
- Appearance:White powder to lump