Order Entry
China
ContactUsLinkComponent
Bis(2,2,2-trifluoroethyl) phthalate, standard for phthalate GLC determination
Bis(2,2,2-trifluoroethyl) phthalate, standard for phthalate GLC determination
Catalog # TCIAP0785-100MG
Supplier:  TCI
CAS Number:  
undefined
Bis(2,2,2-trifluoroethyl) phthalate, standard for phthalate GLC determination
Catalog # TCIAP0785-100MG
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 mg
  • CAS No.:
    62240-27-1
  • Description:
    Bis(2,2,2-trifluoroethyl) Phthalate, ≥98% (T)
  • Formula:
    C₁₂H₈F₆O₄
  • Storage temperature:
    Refrigerator
  • Description:
    Bis(2,2,2-trifluoroethyl) Phthalate
  • Appearance:
    White to Almost white powder to crystal

Specifications

About this item