Order Entry
China
ContactUsLinkComponent
Rhodamine B solution 0,2% in isopropanol (IPA) derivatisation reagent, for TLC, Supelco®
Flammable
Irritant
Rhodamine B solution 0,2% in isopropanol (IPA) derivatisation reagent, for TLC, Supelco®
Catalog # SUPL02558-100ML
Supplier:  SUPELCO
CAS Number:  
undefined
Flammable
Irritant
Rhodamine B solution 0,2% in isopropanol (IPA) derivatisation reagent, for TLC, Supelco®
Catalog # SUPL02558-100ML
Supplier:  SUPELCO
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :479.02 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ100
UN:1219
 :GHS02
 :GHS07
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931