

Cobalt(II) nitrate hexahydrate ≥99.0% ACS, Reag. Ph. Eur. (max.0.001% Ni)
Catalog # VWRC87027.230
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay (Complexometric):99,0 - 102,0
- Identity (IR-Spectrum):passes test
- Insoluble in water:≤ 0,005 %
- Chlorides (Cl):≤ 0,002 %
- Sulfates (SO4):≤ 0,005 %
- Ammonium (NH4):≤ 0,05 %
- Calcium (Ca):≤ 0,005 %
- Copper (Cu):≤ 0,001 %
- Iron (Fe):≤ 0,001 %
- Lead (Pb):≤ 0,001 %
- Magnesium (Mg):≤ 0,005 %
- Manganese (Mn):≤ 0,005 %
- Nickel (Ni):≤ 0,001 %
- Potassium (K):≤ 0,01 %
- Sodium (Na):≤ 0,05 %
- Zinc (Zn):≤ 0,005 %
Specification Test Results
About this item
:
:291.03 g/mol
Merck Index:13,02469
Melting Pt:57 °C
Density:1.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-402-1
UN:1477
ADR:5.1,III
:GHS08
:GHS09
CAS Number:10026-22-9
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647