Order Entry
China
ContactUsLinkComponent
Cobalt(II) nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® ACS analytical reagent
Health
Environment
Cobalt(II) nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® ACS analytical reagent
Catalog # VWRC87024.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Environment
Cobalt(II) nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® ACS analytical reagent
Catalog # VWRC87024.180
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    98.0 --> 102.0 %
  • Insoluble matter:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • NH4 (Ammonium):
    Max. 0.2 %
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Cu (Copper):
    Max. 20 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 100 ppm
  • Mg (Magnesium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 0.05 %
  • Ni (Nickel):
    Max. 0.15 %
  • Pb (Lead):
    Max. 20 ppm
  • Zn (Zinc):
    Max. 100 ppm
  • Conforms to ACS:
    Passes test

Specification Test Results

About this item

 :
 :291.03 g/mol
Merck Index:13,02469
Melting Pt:57 °C
Density:1.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-402-1
UN:1477
ADR:5.1,III
 :GHS08
 :GHS09
CAS Number:10026-22-9
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647