Order Entry
China
ContactUsLinkComponent
Sodium chloride 99.0-100.5% (dry basis), Multi-Compendial, Macron Fine Chemicals™
Sodium chloride 99.0-100.5% (dry basis), Multi-Compendial, Macron Fine Chemicals™
Catalog # MACRH672-06
CAS Number:  
Sodium chloride 99.0-100.5% (dry basis), Multi-Compendial, Macron Fine Chemicals™
Catalog # MACRH672-06
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Acidity or Alkalinity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • Aluminium(Al):
    Max. 0.2 ppm
  • Arsenic (As):
    Max. 1 ppm
  • Barium (Ba):
    Passes Test
  • Limits of Bromides (Br):
    Max. 0.01%
  • Ferrocyanide:
    Passes Test
  • Heavymetals (As Pb):
    Max. 5 ppm
  • Iodides (I):
    Passes Test
  • Iron(Fe):
    Max. 2 ppm
  • Loss On Drying At 105 °C:
    Max. 0.5%
  • Magnesium And Alkaline Earth Metals:
    Max. 0.01%
  • Nitrite (NO₂):
    Max. 0.01%
  • Phosphate (PO₄):
    Max. 0.0025%
  • Potassium (K):
    Max. 0.05%
  • Sulphate(SO₄):
    Max. 0.02%
  • Assay (NaCl) (Dried Basis):
    99.00 to 100.50%
  • Appearence of Solution:
    Passes Test
  • Bromide (Br):
    Max. 100 ppm
  • Iodide(I):
    Passes Test
  • Heavy Metals (As Pb):
    Max. 5 ppm
  • Iron (Fe):
    Max. 2 ppm
  • Potassium(K:
    Max. 500 ppm
  • Endotoxin Conceration:
    Max. 5 IU/g
  • Calarity And Colour of Solution:
    Passes Test
  • Errocyanide:
    Passes Test
  • Iodide (I):
    Passes Test
  • Sulphates (As SO₄):
    Passes Test

Specification Test Results

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477