Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Catalog # BAKR4113-06
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- ≤ 0.10 %:≤100 cfu/g
- Microbiological – Total aerobic microbial count (cfu/g):Diethylene Glycol
- Microbiological – Total yeast and mold count (cfu/g):≤100 cfu/g
- Microbiological - Escherichia Coli:Passes Test
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Residual Solvents - Ethylene Glycol:≤620 ppm
- Residual Solvents - 2-Propanol:≤5000 ppm
- Residual Solvents - Acetic Acid:≤5000 ppm
- NF - Acid Value:≤2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤0.25 %
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25°C:1.06 - 1.09
- NF - Viscosity at 25°C, cSt:300 - 500
- NF - Water (H₂O):≤3.0 %
- NF - Peroxide Value:≤10.0
- NF - Ethylene Oxide:≤1 ppm
- NF - Dioxane:≤10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤5.0 %
- NF - Composition of Fatty Acids - Palmitic Acid:≤16.0 %
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤8.0 %
- NF - Composition of Fatty Acids - Stearic Acid:≤6.0 %
- NF - Composition of Fatty Acids - Oleic Acid:≥58.0 %
- NF - Composition of Fatty Acids - Linoleic Acid:≤18.0 %
- NF - Composition of Fatty Acids - Linolenic Acid:≤4.0 %
- EP - Acid Value:≤2.0
- EP - Total Ash:≤0.25 %
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤2.0
- EP - Ethylene Oxide:≤1 ppm
- EP - Dioxan:≤10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤3.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤5.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤16.0 %
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤8.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤6.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥58.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤18.0 %
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤4.0 %
- JP - Acid Value:≤2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤5.0 %
- JP - Composition of Fatty Acids - Palmitic Acid:≤16.0 %
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤8.0 %
- JP - Composition of Fatty Acids - Stearic Acid:≤6.0 %
- JP - Composition of Fatty Acids - Oleic Acid:≥58.0 %
- JP - Composition of Fatty Acids - Linoleic Acid:≤18.0 %
- JP - Composition of Fatty Acids - Linolenic Acid:≤4.0 %
- JP - Dioxane:≤10 ppm
- JP - Ethylene Oxide:≤1 ppm
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤10.0
- JP - Residue on Ignition:≤0.25 %
- JP - Water (H₂O):≤3.0 %
- JP - Saponification Value:45 - 55
- Appearance (yellow to dark amber, viscous liquid):Passes Test
- Additional Tests - Color (Gardner):≤7
- Additional Tests - Odor (Faint):Passes Test
- Additional Tests - Water (H₂O):≤0.2 %
- Additional Tests - Peroxide Value, meqO₂/kg:≤2.0
- Additional Tests - Endotoxin Concentration (EU/mL):≤10
- Free Ethylene Oxide:≤1 ppm
- 1,4-Dioxane:≤5 ppm
Specification Test Results
About this item
:
:Management of Change (MOC) category = R
Storage Temperature:Refrigerator
CAS Number:9005-65-6
MDL Number:MFCD00082107