Order Entry
China
ContactUsLinkComponent
1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 mol/l (0.1 M), AVS TITRINORM
1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 mol/l (0.1 M), AVS TITRINORM
Catalog # VWRC86187.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 mol/l (0.1 M), AVS TITRINORM
Catalog # VWRC86187.290
Supplier:  VWR Chemicals
CAS Number:  

Specification Test Results

  • Titer (20°C):
    0.9980 – 1.0020
  • Method of analysis::
    complexometric titration
  • Using::
    reference material
  • Indicator::
    eriochrome black T
  • Reference material::
    zinc
  • Traceability::
    the reference material is a secondary reference material,

Specification Test Results

About this item

Synonyms: CDTA disodium salt
Molecular Weight:390.3 g/mol