Supelco® Organic Reference Standard, Pyridoxamine dihydrochloride
SUPLP9380-1G
: SUPELCO
:
- Pk:1 g
- Standards:Analytical standard, ≥98%
- CAS No.:524-36-7
- Description:Pyridoxamine dihydrochloride
- Seq:01
- Storage conditions:–20 °C
- Shipping conditions:STD
- MDL No.:MFCD00012808
- M-Clarity quality level:MQ200
- Synonym(s):4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride
- Melting point:224 - 226 °C (dec.) (lit.)
- Molecular weight:241,11